Alfa Aesar™ Diethyl 5-hydantoylphosphonate, 96%
Manufacturer:  Alfa Aesar™ L1334103
 View more versions of this product
Catalog No. AAL1334103
Specifications
Specifications
Diethyl 5-hydantoylphosphonate
1g
C7H13N2O5P
4809527
PNQYQNIOVAYJJG-UHFFFAOYSA-N
5-diethoxyphosphorylimidazolidine-2,4-dione
3516088
96%
159°C to 161°C
95378-36-2
MFCD00171308
diethyl 2,5-dioxoimidazolidin-4-yl phosphonate, phosphonic acid, 2,5-dioxo-4-imidazolidinyl-, diethyl ester, diethyl 2,5-dioxoimidazolidin-4-ylphosphonate, 5-diethoxyphosphinyl hydantoin, diethyl 2,4-dioxoimidazoline-5-phosphonate, diethyl 2,4-dioxoimidazolidine-5-phosphonate, 5-diethoxycarbonyl-1,3-diazolidine-2,4-dione
CCOP(=O)(C1C(=O)NC(=O)N1)OCC
236.164
236.17
Chemical Identifiers
95378-36-2
236.164
PNQYQNIOVAYJJG-UHFFFAOYSA-N
3516088
CCOP(=O)(C1C(=O)NC(=O)N1)OCC
C7H13N2O5P
MFCD00171308
diethyl 2,5-dioxoimidazolidin-4-yl phosphonate, phosphonic acid, 2,5-dioxo-4-imidazolidinyl-, diethyl ester, diethyl 2,5-dioxoimidazolidin-4-ylphosphonate, 5-diethoxyphosphinyl hydantoin, diethyl 2,4-dioxoimidazoline-5-phosphonate, diethyl 2,4-dioxoimidazolidine-5-phosphonate, 5-diethoxycarbonyl-1,3-diazolidine-2,4-dione
5-diethoxyphosphorylimidazolidine-2,4-dione
Documents