missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Diethyl (5-Bromopentyl)malonate, TCI America™

$203.20 - $524.03

Chemical Identifiers

CAS 1906-95-2
Molecular Formula C12H21BrO4
Molecular Weight (g/mol) 309.2
MDL Number MFCD00048565
InChI Key JZCAZGWJCVMTTQ-UHFFFAOYSA-N
Synonym (5-Bromopentyl)malonic Acid Diethyl Ester
PubChem CID 4401294
IUPAC Name diethyl 2-(5-bromopentyl)propanedioate
SMILES CCOC(=O)C(CCCCCBr)C(=O)OCC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B20835G View Documents TCI America
B20835G
5g Each for $203.20
 
B208325G View Documents TCI America
B208325G
25g Each for $524.03
 
Specifications

Chemical Identifiers

1906-95-2
309.2
JZCAZGWJCVMTTQ-UHFFFAOYSA-N
4401294
CCOC(=O)C(CCCCCBr)C(=O)OCC
C12H21BrO4
MFCD00048565
(5-Bromopentyl)malonic Acid Diethyl Ester
diethyl 2-(5-bromopentyl)propanedioate

Specifications

161°C
5g
C12H21BrO4
(5-Bromopentyl)malonic Acid Diethyl Ester
CCOC(=O)C(CCCCCBr)C(=O)OCC
309.2
309.20
≥97.0% (GC)
Undesignated
1906-95-2
MFCD00048565
JZCAZGWJCVMTTQ-UHFFFAOYSA-N
diethyl 2-(5-bromopentyl)propanedioate
4401294
Clear Liquid at 20°C
Diethyl (5-Bromopentyl)malonate
Safety and Handling

Safety and Handling

TSCA : No