Please login to your online account to display your discounted pricing

Diethyl 5-amino-3-methyl-2,4-thiophenedicarboxylate, 97%, ACROS Organics™


Quantity 50g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  170230500

 View more versions of this product

Catalog No. AC170230500

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥96.0%
Physical Form Crystalline Powder
Formula Weight 257.29
Quantity 50g
Color Slightly yellow to beige-yellow
EINECS Number 225-388-0
Melting Point 103° to 108°C
Chemical Name or Material Diethyl 5-amino-3-methyl-2, 4-thiophenedicarboxylate
Packaging Glass bottle
Name Note 97%
Assay Percent Range 97%
Molecular Formula C11H15NO4S
MDL Number MFCD00005450
CAS 4815-30-9
Infrared Spectrum Authentic

Specifications

Chemical Name or Material Diethyl 5-amino-3-methyl-2, 4-thiophenedicarboxylate
Name Note 97%
CAS 4815-30-9
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C11H15NO4S
Formula Weight 257.29
MDL Number MFCD00005450