Please login to your online account to display your discounted pricing

Diethyl 5-amino-3-methyl-2,4-thiophenedicarboxylate, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Diethyl 5-amino-3-methyl-2, 4-thiophenedicarboxylate
Name Note 97%
CAS 4815-30-9
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C11H15NO4S
Formula Weight 257.29
MDL Number MFCD00005450
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC170230100

  SDS
fisher scientific acros
170230100
Glass bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Crystalline Powder
Assay ≥96.0%
Formula Weight 257.29
Quantity 10g
Color Slightly yellow to beige-yellow
EINECS Number 225-388-0
Melting Point 103° to 108°C
Chemical Name or Material Diethyl 5-amino-3-methyl-2, 4-thiophenedicarboxylate
Packaging Glass bottle
Name Note 97%
Assay Percent Range 97%
Molecular Formula C11H15NO4S
MDL Number MFCD00005450
CAS 4815-30-9
Infrared Spectrum Authentic