Please login to your online account to display your discounted pricing

Diethyl (4-nitrobenzyl)phosphonate, 98%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: fisher scientific acros  332290250

 View more versions of this product

Catalog No. AC332290250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Formula Weight 273.22
Color Clear yellow-orange to dark red
Quantity 25g
Beilstein 16, IV, 1096
Density 1.24g/mL
Chemical Name or Material Diethyl (4-nitrobenzyl)phosphonate
NMR 97.5% min.
Packaging Glass bottle
Refractive Index 1.5220 to 1.5260 (20°C, 589nm)
Assay Percent Range 98%
Molecular Formula C11H16NO5P
MDL Number MFCD00047747
CAS 2609-49-6
Infrared Spectrum Authentic

Specifications

Chemical Name or Material Diethyl (4-nitrobenzyl)phosphonate
Assay Percent Range 98%
Molecular Formula C11H16NO5P
Formula Weight 273.22
MDL Number MFCD00047747