missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Diethyl (4-nitrobenzyl)phosphonate, 98%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 332290050
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
for C-C Bond Formation (Olefination)Specifications
Diethyl (4-nitrobenzyl)phosphonate | |
1.2400g/mL | |
Authentic | |
Glass bottle | |
1.5220 to 1.526 | |
5g | |
1.24 | |
FORMFFDDQMCTCT-UHFFFAOYSA-N | |
1-(diethoxyphosphorylmethyl)-4-nitrobenzene | |
256065 | |
98% |
2609-49-6 | |
151°C (0.1mmHg) | |
98% | |
C11H16NO5P | |
MFCD00047747 | |
16, IV, 1096 | |
diethyl 4-nitrobenzylphosphonate, diethyl 4-nitrobenzyl phosphonate, diethyl 4-nitrophenyl methylphosphonate, 1-diethoxyphosphorylmethyl-4-nitro-benzene, 1-diethoxyphosphorylmethyl-4-nitrobenzene, phosphonic acid, 4-nitrophenyl methyl-, diethyl ester, phosphonic acid, p-4-nitrophenyl methyl-, diethyl ester, zlchem 660, diethyl4-nitrobenzylphosphonate | |
CCOP(=O)(CC1=CC=C(C=C1)[N+](=O)[O-])OCC | |
273.22 | |
273.22 |
Chemical Identifiers
2609-49-6 | |
273.22 | |
FORMFFDDQMCTCT-UHFFFAOYSA-N | |
256065 | |
CCOP(=O)(CC1=CC=C(C=C1)[N+](=O)[O-])OCC |
C11H16NO5P | |
MFCD00047747 | |
diethyl 4-nitrobenzylphosphonate, diethyl 4-nitrobenzyl phosphonate, diethyl 4-nitrophenyl methylphosphonate, 1-diethoxyphosphorylmethyl-4-nitro-benzene, 1-diethoxyphosphorylmethyl-4-nitrobenzene, phosphonic acid, 4-nitrophenyl methyl-, diethyl ester, phosphonic acid, p-4-nitrophenyl methyl-, diethyl ester, zlchem 660, diethyl4-nitrobenzylphosphonate | |
1-(diethoxyphosphorylmethyl)-4-nitrobenzene |
RUO â Research Use Only