Learn More
Diethyl 4-methoxybenzylphosphonate, 98+%, Thermo Scientific Chemicals
$130.82 - $509.71
Chemical Identifiers
CAS | 1145-93-3 |
---|---|
Molecular Formula | C12H19O4P |
Molecular Weight (g/mol) | 258.254 |
MDL Number | MFCD00015662 |
InChI Key | NKARVHNAACNYGE-UHFFFAOYSA-N |
Synonym | diethyl 4-methoxybenzylphosphonate, diethyl 4-methoxybenzyl phosphonate, 1-diethoxyphosphorylmethyl-4-methoxybenzene, diethyl 4-methoxyphenyl methylphosphonate, 1-diethoxyphosphorylmethyl-4-methoxy-benzene, phosphonic acid, 4-methoxyphenyl methyl-, diethyl ester, acmc-1bsuk, 4-diethylphosphonomethyl anisole, diethyl-4-methoxybenzyl phosphonate, diethyl 4-methoxybenzyl-phosphonate |
PubChem CID | 4063826 |
IUPAC Name | 1-(diethoxyphosphorylmethyl)-4-methoxybenzene |
SMILES | CCOP(=O)(CC1=CC=C(C=C1)OC)OCC |
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
AAL0522609
|
Thermo Scientific Chemicals
L0522609 |
10 g |
N/A
|
|
|||||
AAL0522618
|
Thermo Scientific Chemicals
L0522618 |
50 g |
N/A
|
|
|||||
Description
Diethyl 4-methoxybenzylphosphonate is used in synthesis of C-aryl glycosides. It acts as a reactant for synthesis of γ -Monofluorinated goniothalamin analogs via ring-opening hydrofluorination, stilbenoid derivatives with neuroprotective activity, resveratrol-chroman hybrids with antioxidant activity, Oligostilbenoids using regioselective cyclodehydration and arylation.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDiethyl 4-methoxybenzylphosphonate is used in synthesis of C-aryl glycosides. It acts as a reactant for synthesis of γ -Monofluorinated goniothalamin analogs via ring-opening hydrofluorination, stilbenoid derivatives with neuroprotective activity, resveratrol-chroman hybrids with antioxidant activity, Oligostilbenoids using regioselective cyclodehydration and arylation.
Solubility
Not miscible in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible materials are Strong oxidizing agents.
Chemical Identifiers
1145-93-3 | |
258.254 | |
NKARVHNAACNYGE-UHFFFAOYSA-N | |
4063826 | |
CCOP(=O)(CC1=CC=C(C=C1)OC)OCC |
C12H19O4P | |
MFCD00015662 | |
diethyl 4-methoxybenzylphosphonate, diethyl 4-methoxybenzyl phosphonate, 1-diethoxyphosphorylmethyl-4-methoxybenzene, diethyl 4-methoxyphenyl methylphosphonate, 1-diethoxyphosphorylmethyl-4-methoxy-benzene, phosphonic acid, 4-methoxyphenyl methyl-, diethyl ester, acmc-1bsuk, 4-diethylphosphonomethyl anisole, diethyl-4-methoxybenzyl phosphonate, diethyl 4-methoxybenzyl-phosphonate | |
1-(diethoxyphosphorylmethyl)-4-methoxybenzene |
Specifications
1145-93-3 | |
136°C to 138°C (0.7 mmHg) | |
C12H19O4P | |
MFCD00015662 | |
2125148 | |
Not miscible in water. | |
CCOP(=O)(CC1=CC=C(C=C1)OC)OCC | |
258.254 | |
258.25 | |
Diethyl 4-methoxybenzylphosphonate |
1.5 | |
>110°C (230°F) | |
1.5045 | |
10 g | |
diethyl 4-methoxybenzylphosphonate, diethyl 4-methoxybenzyl phosphonate, 1-diethoxyphosphorylmethyl-4-methoxybenzene, diethyl 4-methoxyphenyl methylphosphonate, 1-diethoxyphosphorylmethyl-4-methoxy-benzene, phosphonic acid, 4-methoxyphenyl methyl-, diethyl ester, acmc-1bsuk, 4-diethylphosphonomethyl anisole, diethyl-4-methoxybenzyl phosphonate, diethyl 4-methoxybenzyl-phosphonate | |
NKARVHNAACNYGE-UHFFFAOYSA-N | |
1-(diethoxyphosphorylmethyl)-4-methoxybenzene | |
4063826 | |
≥98% |
Safety and Handling
EINECSNumber : 214-548-5
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only