Please login to your online account to display your discounted pricing

Diethyl (4-chlorobenzyl)phosphonate, 98%, ACROS Organics™


Quantity 5g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  332260050

 View more versions of this product

Catalog No. AC332260050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.5080 to 1.5100 (20°C, 589nm)
Density 1.09g/mL
Color Clear colorless
Quantity 5g
Chemical Name or Material Diethyl (4-chlorobenzyl)phosphonate
CAS 39225-17-7
Physical Form Clear colorless liquid
Assay 97.5% min.
Assay Percent Range 98%
Molecular Formula C11H16ClO3P
Formula Weight 262.67g/mol
MDL Number MFCD00051568
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECS Number 254-365-8

Specifications

Chemical Name or Material Diethyl (4-chlorobenzyl)phosphonate
CAS 39225-17-7
Assay 97.5% min.
Assay Percent Range 98%
Molecular Formula C11H16ClO3P
Formula Weight 262.67g/mol
MDL Number MFCD00051568

for C-C Bond Formation (Olefination)

Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

SDS & Safety Documents