missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Diethyl 4-aminobenzylphosphonate, 98%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 114010050
Description
for C-C Bond Formation (Olefination)Specifications
Diethyl 4-aminobenzylphosphonate | |
Glass bottle | |
90°C to 94°C | |
98% | |
98% | |
(C2H5O)2P(O)CH2C6H4NH2 | |
4-diethoxyphosphorylmethyl aniline, diethyl 4-aminobenzylphosphonate, diethyl 4-aminophenyl methylphosphonate, diethyl p-aminobenzyl phosphonate, diethyl p-aminobenzylphosphonate, p-aminobenzylphosphonic acid diethyl ester, phosphonic acid, 4-aminophenyl methyl-, diethyl ester, phosphonic acid, p-aminobenzyl-, diethyl ester, timtec-bb sbb000830 | |
CCOP(=O)(CC1=CC=C(C=C1)N)OCC | |
243.24 | |
243.24 | |
98% |
Authentic | |
Beige to Brown | |
5g | |
20074-79-7 | |
C11H18NO3P | |
MFCD00007913 | |
ZVAYUUUQOCPZCZ-UHFFFAOYSA-N | |
4-(diethoxyphosphorylmethyl)aniline | |
88358 | |
Crystals or Crystalline Powder |
Chemical Identifiers
20074-79-7 | |
243.24 | |
ZVAYUUUQOCPZCZ-UHFFFAOYSA-N | |
88358 | |
CCOP(=O)(CC1=CC=C(C=C1)N)OCC |
C11H18NO3P | |
MFCD00007913 | |
4-diethoxyphosphorylmethyl aniline, diethyl 4-aminobenzylphosphonate, diethyl 4-aminophenyl methylphosphonate, diethyl p-aminobenzyl phosphonate, diethyl p-aminobenzylphosphonate, p-aminobenzylphosphonic acid diethyl ester, phosphonic acid, 4-aminophenyl methyl-, diethyl ester, phosphonic acid, p-aminobenzyl-, diethyl ester, timtec-bb sbb000830 | |
4-(diethoxyphosphorylmethyl)aniline |
Safety and Handling
EINECSNumber : 243-504-8
RTECSNumber : SZ6480000