Please login to your online account to display your discounted pricing

Diethyl 4-aminobenzylphosphonate 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Diethyl 4-aminobenzylphosphonate
Name Note 98%
CAS 20074-79-7
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C11H18NO3P
Linear Formula (C2H5O)2P(O)CH2C6H4NH2
Formula Weight 243.24
MDL Number MFCD00007913
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC114010050

  SDS
fisher scientific acros
114010050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC114010250

  SDS
fisher scientific acros
114010250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Crystals or Crystalline Powder
Assay ≥97.5%
Formula Weight 243.24
Quantity 5g
Color Beige to light brown
EINECS Number 243-504-8
Melting Point 90° to 94°C
RTECS Number SZ6480000
Packaging Glass bottle
Name Note 98%
MDL Number MFCD00007913
Infrared Spectrum Authentic
Chemical Name or Material Diethyl 4-aminobenzylphosphonate
Assay Percent Range 98%
Molecular Formula C11H18NO3P
Linear Formula (C2H5O)2P(O)CH2C6H4NH2
CAS 20074-79-7

for C-C Bond Formation (Olefination)