missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Diethyl 4,4′-Biphenyldicarboxylate, TCI America™

$163.47 - $484.31

Chemical Identifiers

CAS 47230-38-6
Molecular Formula C18H18O4
Molecular Weight (g/mol) 298.338
MDL Number MFCD00017270
InChI Key SYTZNHBXNLYWAK-UHFFFAOYSA-N
Synonym diethyl 4,4'-biphenyldicarboxylate, diethyl biphenyl-4,4'-dicarboxylate, diethyl 1,1'-biphenyl-4,4'-dicarboxylate, ethyl 4-4-ethoxycarbonylphenyl benzoate, diethyl-4,4'-diphenyldicarboxylate, diethyl biphenyl 4,4'-dicarboxylate, ethyl 4-4-ethoxycarbonyl phenyl benzoate, 4,4'-biphenyldicarboxylic acid diethyl ester, 4,4'-diethyl 1,1'-biphenyl-4,4'-dicarboxylate, acmc-1apwf
PubChem CID 261553
IUPAC Name ethyl 4-(4-ethoxycarbonylphenyl)benzoate
SMILES CCOC(=O)C1=CC=C(C=C1)C2=CC=C(C=C2)C(=O)OCC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
B08311G View Documents TCI America
B08311G
1g Each for $163.47
 
B08315G View Documents TCI America
B08315G
5g Each for $484.31
 
Specifications

Chemical Identifiers

47230-38-6
298.338
SYTZNHBXNLYWAK-UHFFFAOYSA-N
261553
CCOC(=O)C1=CC=C(C=C1)C2=CC=C(C=C2)C(=O)OCC
C18H18O4
MFCD00017270
diethyl 4,4'-biphenyldicarboxylate, diethyl biphenyl-4,4'-dicarboxylate, diethyl 1,1'-biphenyl-4,4'-dicarboxylate, ethyl 4-4-ethoxycarbonylphenyl benzoate, diethyl-4,4'-diphenyldicarboxylate, diethyl biphenyl 4,4'-dicarboxylate, ethyl 4-4-ethoxycarbonyl phenyl benzoate, 4,4'-biphenyldicarboxylic acid diethyl ester, 4,4'-diethyl 1,1'-biphenyl-4,4'-dicarboxylate, acmc-1apwf
ethyl 4-(4-ethoxycarbonylphenyl)benzoate

Specifications

White-Yellow
1g
C18H18O4
diethyl 4,4'-biphenyldicarboxylate, diethyl biphenyl-4,4'-dicarboxylate, diethyl 1,1'-biphenyl-4,4'-dicarboxylate, ethyl 4-4-ethoxycarbonylphenyl benzoate, diethyl-4,4'-diphenyldicarboxylate, diethyl biphenyl 4,4'-dicarboxylate, ethyl 4-4-ethoxycarbonyl phenyl benzoate, 4,4'-biphenyldicarboxylic acid diethyl ester, 4,4'-diethyl 1,1'-biphenyl-4,4'-dicarboxylate, acmc-1apwf
CCOC(=O)C1=CC=C(C=C1)C2=CC=C(C=C2)C(=O)OCC
298.338
298.34
≥98.0% (GC,T)
113°C
47230-38-6
MFCD00017270
SYTZNHBXNLYWAK-UHFFFAOYSA-N
ethyl 4-(4-ethoxycarbonylphenyl)benzoate
261553
Crystal-Powder at 20°C
Diethyl 4,4′-Biphenyldicarboxylate
Safety and Handling

Safety and Handling

TSCA : No