missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Diethyl (3,4-dichlorophenyl)malonate, 95%

$93.05 - $309.66

Chemical Identifiers

CAS 28751-26-0
Molecular Formula C13H14Cl2O4
Formula Weight 305.16
MDL Number MFCD02180874
Synonym (3,4-Dichlorophenyl)malonic acid diethyl ester
Chemical Name or Material Diethyl (3,4-dichlorophenyl)malonate
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAH2627703 SDS Alfa Aesar™
H2627703
1g Each for $93.05
AAH2627706 SDS Alfa Aesar™
H2627706
5g Each for $309.66
Specifications Description & Specifications

Chemical Identifiers

28751-26-0
305.16
(3,4-Dichlorophenyl)malonic acid diethyl ester
C13H14Cl2O4
MFCD02180874
Diethyl (3,4-dichlorophenyl)malonate

Specifications

1g
C13H14Cl2O4
MFCD02180874
Diethyl (3,4-dichlorophenyl)malonate
28751-26-0
305.16
(3,4-Dichlorophenyl)malonic acid diethyl ester
95%

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning