Please login to your online account to display your discounted pricing

Diethyl (2S,3S)-(+)-2,3-epoxysuccinate, 98%, ACROS Organics™


Quantity 100mg
Packaging Glass Bottle in Plastic Container

Manufacturer: thermo fisher scientific  333591000

 View more versions of this product

Catalog No. AC333591000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Liquid
Formula Weight 188.18
Quantity 100mg
Color Clear colorless to slightly yellow
Packaging Glass Bottle in Plastic Container
Specific Rotation Condition + 108.00 (20.00°C c=2,diethylether)
Refractive Index 1.4370 to 1.4390 (20°C, 589nm)
Specific Rotation +104° to +112° (20°C, 589nm) (c=2, diethylether)
Synonym Diethyl (2S-trans)-oxiranedicarboxylate
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material Diethyl (2S, 3S)-(+)-2, 3-epoxysuccinate
Name Note 98%
Assay Percent Range 98%
Molecular Formula C8H12O5
MDL Number MFCD01632600
CAS 73890-18-3
Infrared Spectrum Authentic

Specifications

Chemical Name or Material Diethyl (2S, 3S)-(+)-2, 3-epoxysuccinate
Name Note 98%
CAS 73890-18-3
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C8H12O5
Formula Weight 188.18
MDL Number MFCD01632600
Synonym Diethyl (2S-trans)-oxiranedicarboxylate
Accessories

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.

SDS & Safety Documents