Please login to your online account to display your discounted pricing

Alfa Aesar™ Diethyl (2-pentyl)malonate, 94%

$58.31 - $638.86

Chemical Identifiers

Chemical Name or Material Diethyl (2-pentyl)malonate
CAS 117-47-5
Molecular Formula C12H22O4
Formula Weight 230.31
MDL Number MFCD00051558
Synonym Diethyl sec-amyl malonate; Diethyl (1-methylbutyl)malonate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2499309

  SDS
alfa aesar™
B2499309
10g Each for $58.31

AAB2499318

  SDS
alfa aesar™
B2499318
50g Each for $155.20

AAB2499330

  SDS
alfa aesar™
B2499330
250g Each for $638.86
Description & Specifications

Specifications

Formula Weight 230.31
Chemical Name or Material Diethyl (2-pentyl)malonate
Assay 94%
Molecular Formula C12H22O4
MDL Number MFCD00051558
CAS 117-47-5
Boiling Point 118° to 119°C (9mmHg)
Synonym Diethyl sec-amyl malonate; Diethyl (1-methylbutyl)malonate
Quantity 10g

Applications
Diethyl (2-pentyl)malonate is used in vitro reaction of barbiturates with formaldehyde to evaluate forensic toxicology on the cause of death.

Notes
Incompatible with oxidizing agents.