Please login to your online account to display your discounted pricing

Diethyl 2-isopentyl-2-methylmalonate, Tech., Maybridge

Chemical Identifiers

Chemical Name or Material Diethyl 2-isopentyl-2-methylmalonate
Assay Tech.
Molecular Formula C13H24O4
Formula Weight 244.33
MDL Number MFCD00053035
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

SB00828EA

  SDS
fisher scientific company
SB00828EA
Amber glass bottle 10g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 244.33
Purity Grade Technical Grade
Quantity 10g
Assay Tech.
CAS 121823-85-6
MDL Number MFCD00053035
Packaging Amber glass bottle
Molecular Formula C13H24O4
Chemical Name or Material Diethyl 2-isopentyl-2-methylmalonate