Please login to your online account to display your discounted pricing

Diethyl 2-isopentyl-2-methylmalonate, Tech., Maybridge


Quantity 10g
Packaging Amber glass bottle

Manufacturer: fisher scientific company  SB00828EA

 View more versions of this product

Catalog No. SB00828EA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 244.33
Purity Grade Technical Grade
Quantity 10g
Assay Tech.
CAS 121823-85-6
MDL Number MFCD00053035
Packaging Amber glass bottle
Molecular Formula C13H24O4
Chemical Name or Material Diethyl 2-isopentyl-2-methylmalonate

Specifications

Chemical Name or Material Diethyl 2-isopentyl-2-methylmalonate
Assay Tech.
Molecular Formula C13H24O4
Formula Weight 244.33
MDL Number MFCD00053035