Please login to your online account to display your discounted pricing

Diethyl (2-aminoethyl)phosphonate oxalate, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Diethyl (2-aminoethyl)phosphonate oxalate
CAS 203192-98-7
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C6H16NO3P·C2H2O4
Formula Weight 271.21
MDL Number MFCD00800256
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC332220010

  SDS
fisher scientific acros
332220010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC332220050

  SDS
thermo fisher scientific
AC332220050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Powder or Crystals
Formula Weight 271.21
Quantity 1g
Color White to light yellow
Melting Point 135° to 141°C
Packaging Glass bottle
Health Hazard 1 HYGROSCOPIC
Health Hazard 2 GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Chemical Name or Material Diethyl (2-aminoethyl)phosphonate oxalate
Assay Percent Range 97%
Molecular Formula C6H16NO3P·C2H2O4
MDL Number MFCD00800256
CAS 203192-98-7
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

for C-C Bond Formation (Olefination)

Safety and Handling

HYGROSCOPIC

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.