Please login to your online account to display your discounted pricing

Alfa Aesar™ Diethyl 2-amino-4-methylthiophene-3,5-dicarboxylate, 99%

$179.31 - $626.81

Chemical Identifiers

Chemical Name or Material Diethyl 2-amino-4-methylthiophene-3,5-dicarboxylate
CAS 4815-30-9
Molecular Formula C11H15NO4S
Formula Weight 257.3
MDL Number MFCD00005450
Synonym 2-Amino-3,5-dicarbethoxy-4-methylthiophene; 2-Amino-4-methylthiophene-3,5-dicarboxylic acid diethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1931014

  SDS
alfa aesar™
A1931014
25g Each for $179.31

AAA1931022

  SDS
alfa aesar™
A1931022
100g Each for $626.81
Description & Specifications

Specifications

Formula Weight 257.3
Synonym 2-Amino-3,5-dicarbethoxy-4-methylthiophene; 2-Amino-4-methylthiophene-3,5-dicarboxylic acid diethyl ester
Melting Point 114° to 116°C
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 4815-30-9
Quantity 25g
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 1 Warning
Molecular Formula C11H15NO4S
MDL Number MFCD00005450
Chemical Name or Material Diethyl 2-amino-4-methylthiophene-3,5-dicarboxylate
Assay 99%
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.