Please login to your online account to display your discounted pricing

Alfa Aesar™ Diethyl (2,6-difluorophenyl)malonate, 90+%

$237.24 - $237.24

Chemical Identifiers

Chemical Name or Material Diethyl (2,6-difluorophenyl)malonate
CAS 491860-05-0
Molecular Formula C13H14F2O4
Formula Weight 272.25g/mol
MDL Number MFCD02090536
Synonym (2,6-Difluorophenyl)malonic acid diethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2638527

 
alfa aesar™
H26385-27
2.5g Each for $237.24
Description & Specifications

Specifications

Flash Point >100°C (212°F)
Quantity 2.5g
Chemical Name or Material Diethyl (2,6-difluorophenyl)malonate
CAS 491860-05-0
Assay ≥90%
Molecular Formula C13H14F2O4
Formula Weight 272.25g/mol
MDL Number MFCD02090536
Synonym (2,6-Difluorophenyl)malonic acid diethyl ester
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.