Please login to your online account to display your discounted pricing

Alfa Aesar™ Diethyl (2,5-difluorophenyl)malonate, 95%

$154.69 - $154.69

Chemical Identifiers

Chemical Name or Material Diethyl (2,5-difluorophenyl)malonate
CAS 247170-23-6
Molecular Formula C13H14F2O4
Formula Weight 272.25g/mol
MDL Number MFCD01312759
Synonym (2,5-Difluorophenyl)malonic acid diethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2639306

 
alfa aesar™
H26393-06
5g Each for $154.69
Description & Specifications

Specifications

Quantity 5g
Quantity 5g
Chemical Name or Material Diethyl (2,5-difluorophenyl)malonate
CAS 247170-23-6
Assay 95%
Molecular Formula C13H14F2O4
Formula Weight 272.25g/mol
MDL Number MFCD01312759
Synonym (2,5-Difluorophenyl)malonic acid diethyl ester
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.