Please login to your online account to display your discounted pricing

Diethyl 2,4-dimethylpyrrole-3,5-dicarboxylate, 97%, ACROS Organics™

$155.74 - $155.74

Chemical Identifiers

Chemical Name or Material Diethyl 2, 4-dimethylpyrrole-3, 5-dicarboxylate
Name Note 97%
CAS 2436-79-5
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C12 H17 N O4
Formula Weight 239.27
MDL Number MFCD00005218
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC114280250

  SDS
thermo fisher scientific
114280250
Glass bottle 25g Each for $155.74
Description & Specifications

Specifications

Assay ≥96.0%
Formula Weight 239.27
Physical Form Flaky Powder or Needle-Like Crystals
Quantity 25g
Color Light yellow to light pink
EINECS Number 219-431-2
Melting Point 133° to 138°C
Chemical Name or Material Diethyl 2, 4-dimethylpyrrole-3, 5-dicarboxylate
Packaging Glass bottle
Name Note 97%
MDL Number MFCD00005218
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
Harmful if swallowed.
May cause respiratory irritation.
Harmful in contact with skin.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic
Assay Percent Range 97%
Molecular Formula C12 H17 N O4
CAS 2436-79-5
Safety and Handling

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
Harmful if swallowed.
May cause respiratory irritation.
Harmful in contact with skin.