Please login to your online account to display your discounted pricing

Alfa Aesar™ Diethyl 2,4-dimethylpyrrole-3,5-dicarboxylate, 97%

$34.96 - $121.43

Chemical Identifiers

Chemical Name or Material Diethyl 2,4-dimethylpyrrole-3,5-dicarboxylate
CAS 2436-79-5
Molecular Formula C12H17NO4
Formula Weight 239.27
MDL Number MFCD00005218
Synonym 2,4-Dimethylpyrrole-3,5-dicarboxylic acid diethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2137406

  SDS
alfa aesar™
B2137406
5g Each for $34.96

AAB2137414

  SDS
alfa aesar™
B2137414
25g Each for $121.43
Description & Specifications

Specifications

Formula Weight 239.27
Quantity 5g
Chemical Name or Material Diethyl 2,4-dimethylpyrrole-3,5-dicarboxylate
MDL Number MFCD00005218
CAS 2436-79-5
Molecular Formula C12H17NO4
Melting Point 135° to 138°C
Synonym 2,4-Dimethylpyrrole-3,5-dicarboxylic acid diethyl ester
Assay 97%