Please login to your online account to display your discounted pricing

Alfa Aesar™ Diethyl (2,4,6-trifluorophenyl)malonate, 95%

$158.30 - $158.30

Chemical Identifiers

Chemical Name or Material Diethyl (2,4,6-trifluorophenyl)malonate
CAS 262609-07-4
Molecular Formula C13H13F3O4
Formula Weight 290.24g/mol
MDL Number MFCD04039210
Synonym (2,4,6-Trifluorophenyl)malonic acid diethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH2630403

 
alfa aesar™
H26304-03
1g Each for $158.30
Description & Specifications

Specifications

Melting Point 46° to 49°C
Quantity 1g
Chemical Name or Material Diethyl (2,4,6-trifluorophenyl)malonate
CAS 262609-07-4
Assay 95%
Molecular Formula C13H13F3O4
Formula Weight 290.24g/mol
MDL Number MFCD04039210
Synonym (2,4,6-Trifluorophenyl)malonic acid diethyl ester
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.
Safety and Handling

Warning

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.