missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Diethyl 2,2-dimethoxyethylphosphonate, 97%

$60.66 - $193.95

Chemical Identifiers

CAS 17053-13-3
Molecular Formula C8H19O5P
Formula Weight 226.21
MDL Number MFCD00041388
Beilstein 2253148
Synonym 2,2-Dimethoxyethylphosphonic acid diethyl ester
Chemical Name or Material Diethyl 2,2-dimethoxyethylphosphonate
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL1102504 SDS Alfa Aesar™
L1102504
2g Each for $60.66
AAL1102509 SDS Alfa Aesar™
L1102509
10g Each for $193.95
Specifications Description & Specifications

Chemical Identifiers

17053-13-3
226.21
2253148
Diethyl 2,2-dimethoxyethylphosphonate
C8H19O5P
MFCD00041388
2,2-Dimethoxyethylphosphonic acid diethyl ester

Specifications

2g
C8H19O5P
MFCD00041388
2253148
Diethyl 2,2-dimethoxyethylphosphonate
17053-13-3
226.21
45° to 48°C (0.05mmHg)
2,2-Dimethoxyethylphosphonic acid diethyl ester
97%

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning