missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Diethyl 2,2-dimethoxyethylphosphonate, 97%

Manufacturer:  Alfa Aesar™ L1102509

 View more versions of this product

Catalog No. AAL1102509



Specifications

Specifications

Diethyl 2,2-dimethoxyethylphosphonate
10g
C8H19O5P
2253148
XMORGWRBGODJMM-UHFFFAOYSA-N
2-diethoxyphosphoryl-1,1-dimethoxyethane
3248918
97%
45°C to 48°C (0.05mmHg)
17053-13-3
MFCD00041388
diethyl 2,2-dimethoxyethyl phosphonate, diethyl 2,2-dimethoxyethylphosphonate, acmc-1bt2b, diethylphosphonoacetaldehyde-dimethylacetal, phosphonic acid, 2,2-dimethoxyethyl-, diethyl ester 9ci
CCOP(=O)(CC(OC)OC)OCC
226.209
226.21

Chemical Identifiers

17053-13-3
226.209
XMORGWRBGODJMM-UHFFFAOYSA-N
3248918
CCOP(=O)(CC(OC)OC)OCC
C8H19O5P
MFCD00041388
diethyl 2,2-dimethoxyethyl phosphonate, diethyl 2,2-dimethoxyethylphosphonate, acmc-1bt2b, diethylphosphonoacetaldehyde-dimethylacetal, phosphonic acid, 2,2-dimethoxyethyl-, diethyl ester 9ci
2-diethoxyphosphoryl-1,1-dimethoxyethane
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

Documents