missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Diethyl 2,2-diethoxyethylphosphonate, 96%

Manufacturer:  Alfa Aesar™ A1246109

 View more versions of this product

Catalog No. AAA1246109



Description

Description

Applications
Diethyl 2,2-diethoxyethylphosphonate is used reactant for synthesis of α,β-alkenal derivatives by two-carbon homologation, lower rim-phosphonylated rexorcinol calix[4]arenes by condensation reactions, α-Phosphovinyl radicals via a radical trapping sequence, inhibitors of reverse transcriptase via 1,3-dipolar cycloadditions and for Friedel-Crafts reactions.

Solubility
Soluble in chloroform and dichloromethane. Not miscible or difficult to mix in water.

Notes
Keep container tightly sealed. It is hygroscopic in nature. It is sensitive to moisture. Store under dry inert gas. Protect from humidity and water.
Specifications

Specifications

Diethyl 2,2-diethoxyethylphosphonate
7598-61-0
C10H23O5P
1785614
>110°C (230°F)
LUQYELQXRPNKRY-UHFFFAOYSA-N
2-diethoxyphosphoryl-1,1-diethoxyethane
82071
10g
96%
MFCD00009236
101°C to 103°C (8mmHg)
2,2-Diethoxyethylphosphonic acid diethyl ester; Diethyl phosphonoacetaldehyde diethyl acetal
CCOC(CP(=O)(OCC)OCC)OCC
254.263
254.26

Chemical Identifiers

7598-61-0
254.263
LUQYELQXRPNKRY-UHFFFAOYSA-N
82071
CCOC(CP(=O)(OCC)OCC)OCC
C10H23O5P
MFCD00009236
2,2-Diethoxyethylphosphonic acid diethyl ester; Diethyl phosphonoacetaldehyde diethyl acetal
2-diethoxyphosphoryl-1,1-diethoxyethane
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P280-P305+P351+P338-P362-P321-P332+P313-P337+P313
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Take off contaminated clothing and wash before reuse.
Specific treatment (see label).
If skin irritation occurs: Get medical advice/attention.
If eye irritation persists: Get medical advice/attention.

Warning

Recommended Storage : 2°C to 8°C

SDS Documents