Please login to your online account to display your discounted pricing

Diethyl 2-(2-cyanoethyl)malonate, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Diethyl 2-(2-cyanoethyl)malonate
Name Note 98%
CAS 17216-62-5
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C10 H15 N O4
Linear Formula NCCH2CH2CH(CO2C2H5)2
Formula Weight 213.23
MDL Number MFCD00001966
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC170420050

  SDS
thermo fisher scientific
170420050
Glass bottle 5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥97.5%
Physical Form Liquid
Formula Weight 213.23
Quantity 5g
Color Clear colorless to light yellow
EINECS Number 241-260-7
Density 1.0700g/mL
Refractive Index 1.4350 to 1.4370 (20°C, 589nm)
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.
Health Hazard 1 GHS Signal Word: Warning
Packaging Glass bottle
Infrared Spectrum Authentic
CAS 17216-62-5
MDL Number MFCD00001966
Name Note 98%
Assay Percent Range 98%
Chemical Name or Material Diethyl 2-(2-cyanoethyl)malonate
Molecular Formula C10 H15 N O4
Linear Formula NCCH2CH2CH(CO2C2H5)2
Safety and Handling

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful in contact with skin.
Harmful if inhaled.