missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Diethyl 2-(2-cyanoethyl)malonate, 98+%

Manufacturer:  Alfa Aesar™ A1156718

 View more versions of this product

Catalog No. AAA1156718


 

Specifications

Specifications

Diethyl 2- (2-cyanoethyl)malonate
17216-62-5
C10H15NO4
UN3276
115°C to 117°C (2mmHg)
4,4-Bis(ethoxycarbonyl)butyronitrile; 2-Cyanoethylmalonic acid diethyl ester
CCOC(=O)C(CCC#N)C(=O)OCC
213.233
213.23
50g
≥98%
MFCD00001966
1790237
>110°C (230°F)
YJJLOESDBPRZIP-UHFFFAOYSA-N
diethyl 2-(2-cyanoethyl)propanedioate
87005

Chemical Identifiers

17216-62-5
213.233
YJJLOESDBPRZIP-UHFFFAOYSA-N
87005
C10H15NO4
MFCD00001966
4,4-Bis(ethoxycarbonyl)butyronitrile; 2-Cyanoethylmalonic acid diethyl ester
Safety and Handling

Safety and Handling

GHS H Statement
H331-H302-H312
Toxic if inhaled.
Harmful if swallowed.
Harmful in contact with skin.

GHS P Statement
P261-P280-P304+P340-P311-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/physician.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, LIQUID, TOXIC, N.O.S.

SDS Documents