missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Diethyl 1-propanephosphonate, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 35590-0250

 View more versions of this product

Catalog No. AC355900250

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥96.0%
180.19
Clear colorless
1.4190 to 1.4210 (20°C, 589nm)
97%
MFCD00015139
Liquid
25g
Authentic
Diethyl 1-propanephosphonate
C7H17O3P
18812-51-6

Chemical Identifiers

18812-51-6
C7H17O3P
MFCD00015139
≥96.0%
97%
180.19
Diethyl 1-propanephosphonate