Please login to your online account to display your discounted pricing

Diethyl 1-propanephosphonate, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Diethyl 1-propanephosphonate
CAS 18812-51-6
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C7H17O3P
Formula Weight 180.19
MDL Number MFCD00015139
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC355900250

  SDS
thermo fisher scientific
35590-0250
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Liquid
Formula Weight 180.19
Quantity 25g
Color Clear colorless
Infrared Spectrum Authentic
Refractive Index 1.4190 to 1.4210 (20°C, 589nm)
Chemical Name or Material Diethyl 1-propanephosphonate
Assay Percent Range 97%
Molecular Formula C7H17O3P
MDL Number MFCD00015139
CAS 18812-51-6

for C-C Bond Formation (Olefination)