Please login to your online account to display your discounted pricing

Alfa Aesar™ Diethyl 1-naphthylmethylphosphonate, 95%

$147.51 - $599.69

Chemical Identifiers

Chemical Name or Material Diethyl 1-naphthylmethylphosphonate
CAS 53575-08-9
Molecular Formula C15H19O3P
Formula Weight 278.29
MDL Number MFCD00015769
Beilstein 2943656
Synonym 1-Naphthylmethylphosphonic acid diethyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0528609

  SDS
alfa aesar™
L0528609
10g Each for $147.51

AAL0528618

  SDS
alfa aesar™
L0528618
50g Each for $599.69
Description & Specifications

Specifications

Formula Weight 278.29
Chemical Name or Material Diethyl 1-naphthylmethylphosphonate
Assay 95%
Molecular Formula C15H19O3P
MDL Number MFCD00015769
CAS 53575-08-9
Boiling Point 205° to 206°C (5mmHg)
Beilstein 2943656
Synonym 1-Naphthylmethylphosphonic acid diethyl ester
RTECS Number SZ9852000
Quantity 10g