missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Diethyl 1-decylphosphonate, 97%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L1303806

 View more versions of this product

Catalog No. AAL1303806



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Diethyl 1-decylphosphonate
128°C to 130°C (0.2 mmHg)
1.439
5 g
diethyl 1-decylphosphonate, diethyl decylphosphonate, decylphosphonic acid diethyl ester, decyldiethoxyphosphino-1-one, acmc-20al9l, diethyl n-decanephosphonate, diethyl 1-decanephosphonate, diethyl 1-decyl phosphonate, decyl-phosphonic acid diethyl ester, n-decylphosphonic acid diethyl ester
CCCCCCCCCCP(=O)(OCC)OCC
278.373
278.38
16165-68-7
C14H31O3P
MFCD00015593
1784261
HYQXIVAZQBYBAF-UHFFFAOYSA-N
1-diethoxyphosphoryldecane
2773697
97%

Chemical Identifiers

16165-68-7
278.373
HYQXIVAZQBYBAF-UHFFFAOYSA-N
2773697
CCCCCCCCCCP(=O)(OCC)OCC
C14H31O3P
MFCD00015593
diethyl 1-decylphosphonate, diethyl decylphosphonate, decylphosphonic acid diethyl ester, decyldiethoxyphosphino-1-one, acmc-20al9l, diethyl n-decanephosphonate, diethyl 1-decanephosphonate, diethyl 1-decyl phosphonate, decyl-phosphonic acid diethyl ester, n-decylphosphonic acid diethyl ester
1-diethoxyphosphoryldecane
Safety and Handling

Safety and Handling

TSCA : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only