Please login to your online account to display your discounted pricing

Alfa Aesar™ Diethyl 1-butylphosphonate, 99+%

$89.80 - $292.31

Chemical Identifiers

Chemical Name or Material Diethyl 1-butylphosphonate
CAS 2404-75-3
Molecular Formula C8H19O3P
Formula Weight 194.21
MDL Number MFCD00082700
Beilstein 1763851
Synonym 1-Butylphosphonic acid diethyl ester; Diethyl 1-butanephosphonate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2399706

  SDS
alfa aesar™
B2399706
5g Each for $89.80

AAB2399714

  SDS
alfa aesar™
B2399714
25g Each for $292.31
Description & Specifications

Specifications

Formula Weight 194.21
Boiling Point 74° to 76°C (1mmHg)
Synonym 1-Butylphosphonic acid diethyl ester; Diethyl 1-butanephosphonate
Odor Odorless
Quantity 5g
Assay ≥99%
Molecular Formula C8H19O3P
MDL Number MFCD00082700
CAS 2404-75-3
Chemical Name or Material Diethyl 1-butylphosphonate
Beilstein 1763851