missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Diethyl 1,1-cyclopropanedicarboxylate, 97%, ACROS Organics™

Manufacturer:  Fisher Scientific Acros 164450250

 View more versions of this product

Catalog No. AC164450250

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

Diethyl 1, 1-cyclopropanedicarboxylate
86°C
Glass bottle
1.05
Colorless
97%
96% min. (GC)
C3H4(CO2C2H5)2
1,1-bis ethoxycarbonyl cyclopropane
CCOC(=O)C1(CC1)C(=O)OCC
186.21
186.21
97%
1.0500g/mL
Authentic
1.4320 to 1.434
94°C to 96°C (10.0mmHg)
25g
1559-02-0
C9H14O4
MFCD00001283
KYYUCZOHNYSLFV-UHFFFAOYSA-N
diethyl cyclopropane-1,1-dicarboxylate
73790
Liquid

Chemical Identifiers

1559-02-0
186.21
KYYUCZOHNYSLFV-UHFFFAOYSA-N
73790
CCOC(=O)C1(CC1)C(=O)OCC
C9H14O4
MFCD00001283
1,1-bis ethoxycarbonyl cyclopropane
diethyl cyclopropane-1,1-dicarboxylate
Safety and Handling

Safety and Handling

EINECSNumber : 216-321-6

SDS Documents