Please login to your online account to display your discounted pricing

Diethyl 1,1-cyclopropanedicarboxylate, 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material Diethyl 1, 1-cyclopropanedicarboxylate
Name Note 97%
CAS 1559-02-0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C9H14O4
Linear Formula C3H4(CO2C2H5)2
Formula Weight 186.21
MDL Number MFCD00001283
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC164450250

  SDS
fisher scientific acros
164450250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 186.21
Assay ≥96.0%
Physical Form Liquid
Quantity 25g
Color Clear colorless
EINECS Number 216-321-6
Density 1.0500g/mL
Refractive Index 1.4320 to 1.4340 (20°C, 589nm)
Packaging Glass bottle
Infrared Spectrum Authentic
Name Note 97%
Chemical Name or Material Diethyl 1, 1-cyclopropanedicarboxylate
MDL Number MFCD00001283
Molecular Formula C9H14O4
CAS 1559-02-0
Linear Formula C3H4(CO2C2H5)2
Assay Percent Range 97%