Learn More
Diethoxymethylsilane, 96%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1015318
Description
Diethoxymethylsilane is used in the preparation of N,N'-methylsilanediyl-bis-phthalimide by reacting with phthalimide. It is used as a reagent in the selective reduction of carbonyl compounds, hydorosilylation of alkenes, tandem reductive aldol reaction and rhodium-catalyszed silylcarbocyclization.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsDiethoxymethylsilane is used in the preparation of N,N′-methylsilanediyl-bis-phthalimide by reacting with phthalimide. It is used as a reagent in the selective reduction of carbonyl compounds, hydorosilylation of alkenes, tandem reductive aldol reaction and rhodium-catalyszed silylcarbocyclization.
Solubility
Miscible with water, 1,4-dioxane, tetrahydrofuran, acetonitrile, dichloromethane, chloroform, dimethyl sulfoxide, hexamethylphosphoric triamide and toluene. Insoluble in diethyl ether and hexane.
Notes
Incompatible with oxidizing agents, acids and bases.
Specifications
94°C to 95°C | |
2031-62-1 | |
C5H13O2Si | |
MFCD00026749 | |
1733975 | |
diethoxymethylsilane, methyldiethoxysilane, diethoxy methyl silane, silane, diethoxymethyl, diethoxy-methylsilane, diethoxy methyl silicon, diethoxymethylsilane. methylhydrogendiethoxysilane., diethoxymethyl-silan, diethoxy methyl silyl | |
GAURFLBIDLSLQU-UHFFFAOYSA-N | |
diethoxy(methyl)silicon | |
6327387 | |
96% | |
Moisture sensitive |
Diethoxymethylsilane | |
50 g | |
CH3SiH(OCH2CH3)2 | |
UN1993 | |
10°C (50°F) | |
Miscible with water,1,4-dioxane,tetrahydrofuran,acetonitrile,dichloromethane,chloroform,dimethyl sulfoxide,hexamethylphosphoric triamide and toluene. Insoluble in diethyl ether and hexane. | |
CCO[Si](C)OCC | |
133.242 | |
134.25 | |
1.376 | |
0.84 |
Chemical Identifiers
2031-62-1 | |
133.242 | |
GAURFLBIDLSLQU-UHFFFAOYSA-N | |
6327387 | |
CCO[Si](C)OCC |
C5H13O2Si | |
MFCD00026749 | |
diethoxymethylsilane, methyldiethoxysilane, diethoxy methyl silane, silane, diethoxymethyl, diethoxy-methylsilane, diethoxy methyl silicon, diethoxymethylsilane. methylhydrogendiethoxysilane., diethoxymethyl-silan, diethoxy methyl silyl | |
diethoxy(methyl)silicon |
Safety and Handling
GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
H225-H315-H319-H335
DOTInformation : Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber : 217-982-3
TSCA : Yes
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only