missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Diethoxymethyleacetate, 96%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC171821000

 View more versions of this product

Catalog No. AC171821000

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

Diethoxymethyleacetate
14036-06-7
Authentic
1.3980 to 1.4000 (20°C, 589nm)
≥95%
C7H14O4
162.19
100g
0.99g/mL
Glass bottle
Liquid
96%
CH3CO2CH(OC2H5)2
Clear colorless

Chemical Identifiers

96%
CH3CO2CH(OC2H5)2
Diethoxymethyleacetate
C7H14O4
162.19
≥95%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning