missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Diethoxymethyleacetate, 96%, ACROS Organics™

Chemical Identifiers

Assay Percent Range 96%
Molecular Formula C7H14O4
Linear Formula CH3CO2CH(OC2H5)2
Formula Weight 162.19
Chemical Name or Material Diethoxymethyleacetate
Assay ≥95%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC171820050 SDS Thermo Fisher Scientific
AC171820050
Glass bottle 5g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC171820250 SDS Thermo Fisher Scientific
AC171820250
Glass bottle 25g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC171821000 SDS Thermo Fisher Scientific
AC171821000
Glass bottle 100g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

96%
CH3CO2CH(OC2H5)2
Diethoxymethyleacetate
C7H14O4
162.19
≥95%

Specifications

Liquid
5g
0.99g/mL
CH3CO2CH(OC2H5)2
C7H14O4
14036-06-7
Diethoxymethyleacetate
162.19
Clear colorless
Glass bottle
1.3980 to 1.4000 (20°C, 589nm)
96%
Authentic
≥95%

Safety and Handling

GHS H Statement:
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning