Please login to your online account to display your discounted pricing

Diethoxymethyleacetate, 96%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC171820250

 View more versions of this product

Catalog No. AC171820250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Liquid
Formula Weight 162.19
Assay ≥95%
Quantity 25g
Color Clear colorless
Density 0.99g/mL
Linear Formula CH3CO2CH(OC2H5)2
Refractive Index 1.3980 to 1.4000 (20°C, 589nm)
Chemical Name or Material Diethoxymethyleacetate
Assay Percent Range 96%
Molecular Formula C7H14O4
CAS 14036-06-7
Packaging Glass bottle
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 1 GHS Signal Word: Warning

Specifications

Chemical Name or Material Diethoxymethyleacetate
Assay ≥95%
Assay Percent Range 96%
Molecular Formula C7H14O4
Linear Formula CH3CO2CH(OC2H5)2
Formula Weight 162.19
Accessories

GHS Signal Word: Warning

GHS P Statement:
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.

SDS & Safety Documents