missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Diethoxymethane, 99%, pure, stabilized, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 296450050

 View more versions of this product

Catalog No. AC296450050

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

≥98.5%
Extra Pure
5mL
0.831g/mL
0.02% max. (K.F.)
Authentic
Glass bottle
10ppm max.
99%
CH3CH2OCH2OCH2CH3
Diethoxymethane
Liquid
104.15
Clear colorless
462-95-3,128-37-0
100ppm BHT max.
Ethylal
1.3720 to 1.3740 (20°C, 589nm)
99%
C5H12O2
MFCD00009246

Chemical Identifiers

462-95-3,128-37-0
C5H12O2
104.15
Ethylal
≥98.5%
99%
CH3CH2OCH2OCH2CH3
MFCD00009246
Diethoxymethane, 99%
Extra Pure

Safety and Handling

SDS & Safety Documents

GHS H Statement:
May cause respiratory irritation.
Causes serious eye irritation.
Causes skin irritation.
Highly flammable liquid and vapor.
Toxic in contact with skin.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Gently wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 207-330-6

RTECSNumber : PA8500000

TSCA : TSCA