missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Diethanolamine, 99%, ACROS Organics™
Manufacturer: Fisher Scientific Acros 113920025
Specifications
Diethanolamine | |
179°C | |
Glass Bottle | |
2hPa at 10°C | |
268.0°C | |
27.0°C to 30.0°C | |
111-42-2 | |
C4H11NO2 | |
MFCD00002843 | |
15, 3118 | |
ZBCBWPMODOFKDW-UHFFFAOYSA-N | |
2-(2-hydroxyethylamino)ethanol | |
8113 | |
105.14 | |
99% |
1.0955g/mL | |
Authentic | |
1.4750 to 1.4790 | |
0.2% max. (K.F.) | |
Undesignated | |
2.5kg | |
99% | |
(HOCH2CH2)2NH | |
04, 283 | |
2,2'-dihydroxydiethylamine, 2,2'-iminodiethanol, bis 2-hydroxyethyl amine, diethanolamin, diethanolamine, diethylolamine, diolamine, ethanol, 2,2'-iminobis, iminodiethanol, n,n-diethanolamine | |
C(CO)NCCO | |
105.137 | |
CHEBI:28123 | |
Viscous Liquid or Low Melting Solid |
Chemical Identifiers
111-42-2 | |
105.137 | |
ZBCBWPMODOFKDW-UHFFFAOYSA-N | |
8113 | |
2-(2-hydroxyethylamino)ethanol |
C4H11NO2 | |
MFCD00002843 | |
2,2'-dihydroxydiethylamine, 2,2'-iminodiethanol, bis 2-hydroxyethyl amine, diethanolamin, diethanolamine, diethylolamine, diolamine, ethanol, 2,2'-iminobis, iminodiethanol, n,n-diethanolamine | |
CHEBI:28123 | |
C(CO)NCCO |
Safety and Handling
GHS P Statement
Causes serious eye damage.
Harmful if swallowed.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
Emergency Overview
Harmful if swallowed. Irritating to eyes and skin. Risk of serious damage to eyes. May cause irritation of respiratory tract. Corrosive to metals.
NFPA&C
Danger