Learn More
Diethanolamine, 99%, Thermo Scientific Chemicals
Supplier: Thermo Fisher Scientific 113920010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Diethanolamine | |
1.0955g/mL | |
99% | |
Authentic | |
MFCD00002843 | |
04, 283 | |
1.4760 to 1.4790 (20°C, 589nm) | |
2,2'-dihydroxydiethylamine, 2,2'-iminodiethanol, bis 2-hydroxyethyl amine, diethanolamin, diethanolamine, diethylolamine, diolamine, ethanol, 2,2'-iminobis, iminodiethanol, n,n-diethanolamine | |
OCCNCCO | |
105.14 | |
CHEBI:28123 | |
Viscous Liquid or Low Melting Solid | |
2hPa at 10°C |
111-42-2 | |
Undesignated | |
C4H11NO2 | |
(HOCH2CH2)2NH | |
Glass Bottle | |
15, 3118 | |
1kg | |
ZBCBWPMODOFKDW-UHFFFAOYSA-N | |
2-(2-hydroxyethylamino)ethanol | |
8113 | |
105.14 | |
≥98.5% | |
0.2% max. (K.F.) |
Chemical Identifiers
111-42-2 | |
105.14 | |
ZBCBWPMODOFKDW-UHFFFAOYSA-N | |
8113 | |
2-(2-hydroxyethylamino)ethanol |
C4H11NO2 | |
MFCD00002843 | |
2,2'-dihydroxydiethylamine, 2,2'-iminodiethanol, bis 2-hydroxyethyl amine, diethanolamin, diethanolamine, diethylolamine, diolamine, ethanol, 2,2'-iminobis, iminodiethanol, n,n-diethanolamine | |
CHEBI:28123 | |
OCCNCCO |
Safety and Handling
GHS P Statement
Causes serious eye damage.
Harmful if swallowed.
May cause damage to organs through prolonged or repeated exposure.
Causes skin irritation.
Emergency Overview
Harmful if swallowed. Irritating to eyes and skin. Risk of serious damage to eyes. May cause irritation of respiratory tract. Corrosive to metals.
NFPA&C
Danger
RUO – Research Use Only