Please login to your online account to display your discounted pricing

Alfa Aesar™ Dicyclopropyl ketone, 96%

$61.26 - $61.26

Chemical Identifiers

Chemical Name or Material Dicyclopropyl ketone
CAS 1121-37-5
Molecular Formula C7H10O
Formula Weight 110.16
MDL Number MFCD00001276
UN Number UN1224
Beilstein 774172
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1158014

  SDS
alfa aesar™
A1158014
25g Each for $61.26

AAA1158022

  SDS
alfa aesar™
A1158022
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 110.16
Health Hazard 3 GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Chemical Name or Material Dicyclopropyl ketone
Assay 96%
Molecular Formula C7H10O
MDL Number MFCD00001276
CAS 1121-37-5
UN Number UN1224
Boiling Point 160° to 162°C
Beilstein 774172
Flash Point 39°C (102°F)
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H301-H318-H226-H315-H335
Toxic if swallowed.
Causes serious eye damage.
Flammable liquid and vapor.
Causes skin irritation.
May cause respiratory irritation.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KETONES, LIQUID, N.O.S.
Quantity 25g

Applications
Dicyclopropyl ketone is used as catalytic agent and petrochemical additive. It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Soluble in water.

Notes
Store in a cool, dry, well-ventilated area away from incompatible substances such as oxidizing agents.

Safety and Handling

Danger

GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H318-H226-H315-H335
Toxic if swallowed.
Causes serious eye damage.
Flammable liquid and vapor.
Causes skin irritation.
May cause respiratory irritation.