missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Dicyclopropyl ketone, 96%

Manufacturer:  Alfa Aesar™ A1158022

 View more versions of this product

Catalog No. AAA1158022



Description

Description

Applications
Dicyclopropyl ketone is used as catalytic agent and petrochemical additive. It can be used in agrochemical, pharmaceutical and dyestuff field.

Solubility
Soluble in water.

Notes
Store in a cool, dry, well-ventilated area away from incompatible substances such as oxidizing agents.
Specifications

Specifications

Dicyclopropyl ketone
160°C to 162°C
1121-37-5
MFCD00001276
774172
BIPUHAHGLJKIPK-UHFFFAOYSA-N
dicyclopropylmethanone
70721
96%
39°C (102°F)
100g
C7H10O
UN1224
dicyclopropyl ketone, cyclopropyl ketone, dicyclopropylketone, methanone, dicyclopropyl, dicyclopropyl-methanone, dicyclopropylmethyloxime, dicyclopropyl methyl ketone, pubchem13596, methanone,dicyclopropyl, acmc-1busz
C1CC1C(=O)C2CC2
110.156
110.16

Chemical Identifiers

1121-37-5
110.156
BIPUHAHGLJKIPK-UHFFFAOYSA-N
70721
C1CC1C(=O)C2CC2
C7H10O
MFCD00001276
dicyclopropyl ketone, cyclopropyl ketone, dicyclopropylketone, methanone, dicyclopropyl, dicyclopropyl-methanone, dicyclopropylmethyloxime, dicyclopropyl methyl ketone, pubchem13596, methanone,dicyclopropyl, acmc-1busz
dicyclopropylmethanone
Safety and Handling

Safety and Handling

GHS H Statement
H301-H318-H226-H315-H335
Toxic if swallowed.
Causes serious eye damage.
Flammable liquid and vapor.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement
P210-P301+P310-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: KETONES, LIQUID, N.O.S.

SDS Documents