missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Dicyclopentenyl acrylate

Manufacturer:  Alfa Aesar™ 4686414

 View more versions of this product

Catalog No. AA4686414



Specifications

Specifications

Dicyclopentenyl acrylate
33791-58-1
C13H17O2
Brown
131°C (268°F)
205.28
Liquid
25g
1.07g/mL
MFCD00171305
-40°C
DCPA; Tricyclo[5.2.1.02.6]dec-4-enyl acrylate
204.27

Chemical Identifiers

33791-58-1
205.28
DCPA; Tricyclo[5.2.1.02.6]dec-4-enyl acrylate
C13H17O2
MFCD00171305
Safety and Handling

Safety and Handling

GHS H Statement
H334-H335-H315-H319-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.

GHS P Statement
P284a-P261-P280-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

TSCA : TSCA

Recommended Storage : 2°C to 8°C

SDS Documents