missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Dicyclopentanylmethyl acrylate

Manufacturer:  Alfa Aesar™ 4665222

 View more versions of this product

Catalog No. AA4665222



Specifications

Specifications

Dicyclopentanylmethyl acrylate
100g
C14H20O2
octahydro-4,7-methano-1h-indenyl methyl acrylate, acmc-20mslm, tricyclo 5.2.1.0 2 ,? decan-3-ylmethyl prop-2-enoate
C=CC(=O)OCC1CCC2C1C3CCC2C3
22051805
Liquid
Yellow
93962-84-6
MFCD22380704
PMPNHSZLJPXGCD-UHFFFAOYSA-N
220.312
220.31

Chemical Identifiers

93962-84-6
220.312
PMPNHSZLJPXGCD-UHFFFAOYSA-N
22051805
C14H20O2
MFCD22380704
octahydro-4,7-methano-1h-indenyl methyl acrylate, acmc-20mslm, tricyclo 5.2.1.0 2 ,? decan-3-ylmethyl prop-2-enoate
C=CC(=O)OCC1CCC2C1C3CCC2C3
Safety and Handling

Safety and Handling

GHS H Statement
H334-H335-H315-H319-H317
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
May cause respiratory irritation.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.

GHS P Statement
P284a-P261-P280-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

Recommended Storage : 2°C to 8°C

SDS Documents