missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Dicyclopentadiene, typically 95%, stab.

Manufacturer:  Alfa Aesar™ 3254630

 View more versions of this product

Catalog No. AA3254630



Specifications

Specifications

Dicyclopentadiene
26° (78°F)
171°C to 173°C
Stabilized
C10H12
UN2048
14,2739
HECLRDQVFMWTQS-UHFFFAOYSA-N
132.206
CHEBI:34695
Liquid
0.982g/mL
1.5100
250g
77-73-6
MFCD00078246
1904092
dicyclopentadiene, biscyclopentadiene, cyclopentadiene dimer, bicyclopentadiene, dicyklopentadien, dimer cyklopentadienu, dcpd, 4,7-methano-1h-indene, 3a,4,7,7a-tetrahydro, 3a,4,7,7a-tetrahydro-1h-4,7-methanoindene, 1,3-cyclopentadiene, dimer
C1C=CC2C1C3CC2C=C3
6492
132.21
95% (typically)

Chemical Identifiers

77-73-6
132.206
HECLRDQVFMWTQS-UHFFFAOYSA-N
6492
C1C=CC2C1C3CC2C=C3
C10H12
MFCD00078246
dicyclopentadiene, biscyclopentadiene, cyclopentadiene dimer, bicyclopentadiene, dicyklopentadien, dimer cyklopentadienu, dcpd, 4,7-methano-1h-indene, 3a,4,7,7a-tetrahydro, 3a,4,7,7a-tetrahydro-1h-4,7-methanoindene, 1,3-cyclopentadiene, dimer
CHEBI:34695
Safety and Handling

Safety and Handling

GHS H Statement
H225-H302-H332-H315-H319-H335
Highly flammable liquid and vapor.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P261-P303+P361+P353-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: DICYCLOPENTADIENE

RTECSNumber : PC1050000

TSCA : TSCA

SDS Documents