Please login to your online account to display your discounted pricing

Alfa Aesar™ Dicyclohexyltin dibromide

Chemical Identifiers

Chemical Name or Material Dicyclohexyltin dibromide
CAS 2954-94-1
Molecular Formula C12H22Br2Sn
Formula Weight 444.82
MDL Number MFCD00015370
UN Number UN3146
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA8780003

  SDS
alfa aesar™
8780003
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA8780006

  SDS
alfa aesar™
8780006
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 444.82
Health Hazard 3 GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
UN Number UN3146
Quantity 1g
Chemical Name or Material Dicyclohexyltin dibromide
Melting Point 58° to 64°C
Health Hazard 2 GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Color White
Physical Form Crystalline
MDL Number MFCD00015370
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, SOLID, N.O.S.
Health Hazard 1 Danger
Sensitivity Moisture Sensitive
CAS 2954-94-1
Boiling Point ≈150°C (0.3mmHg)
Molecular Formula C12H22Br2Sn
Safety and Handling

Danger

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.