missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Dicyclohexyltin dibromide

Manufacturer:  Alfa Aesar™ 8780006

 View more versions of this product

Catalog No. AA8780006



Specifications

Specifications

Dicyclohexyltin dibromide
58°C to 64°C
5g
Moisture Sensitive
MFCD00015370
dicyclohexyltin dibromide, stannane, dibromodicyclohexyl, dibromo dicyclohexyl stannane, dicyclohexyltindibromide, stannane,dibromodicyclohexyl
C1CCC(CC1)[Sn](C2CCCCC2)(Br)Br
444.826
444.82
~150°C (0.3mmHg)
2954-94-1
White
C12H22Br2Sn
UN3146
RHFTTZVNBAHQBF-UHFFFAOYSA-L
dibromo(dicyclohexyl)stannane
97477
Crystalline

Chemical Identifiers

2954-94-1
444.826
RHFTTZVNBAHQBF-UHFFFAOYSA-L
97477
C1CCC(CC1)[Sn](C2CCCCC2)(Br)Br
C12H22Br2Sn
MFCD00015370
dicyclohexyltin dibromide, stannane, dibromodicyclohexyl, dibromo dicyclohexyl stannane, dicyclohexyltindibromide, stannane,dibromodicyclohexyl
dibromo(dicyclohexyl)stannane
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P301+P310-P305+P351+P338-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, SOLID, N.O.S.

SDS Documents