Please login to your online account to display your discounted pricing

Alfa Aesar™ Dicyclohexylmethanol, 98%

$38.49 - $467.95

Chemical Identifiers

Chemical Name or Material Dicyclohexylmethanol
CAS 4453-82-1
Molecular Formula C13H24O
Formula Weight 196.33
MDL Number MFCD00001474
Beilstein 1926669
Synonym Dicyclohexyl carbinol
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2009903

  SDS
alfa aesar™
B2009903
1g Each for $38.49

AAB2009906

  SDS
alfa aesar™
B2009906
5g Each for $111.91

AAB2009914

  SDS
alfa aesar™
B2009914
25g Each for $467.95
Description & Specifications

Specifications

Formula Weight 196.33
Chemical Name or Material Dicyclohexylmethanol
Assay 98%
Molecular Formula C13H24O
MDL Number MFCD00001474
CAS 4453-82-1
Boiling Point 153° to 155°C (12mmHg)
Beilstein 1926669
Melting Point 58° to 64°C
Synonym Dicyclohexyl carbinol
Quantity 1g